ENA372573755 Enamine

3,5-dichloro-N-methylpyridin-2-amine

  • MDL number MFCD09745416
  • Cas Number : 4088-62-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113479

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113479
Availability details for
Quantity
  • Availability
  • Add to Cart