ENA372573759 Enamine

5-(5-chlorothiophen-2-yl)-1,2-oxazole-4-carboxylic acid

  • MDL number MFCD21888796
  • Cas Number : 1368489-92-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112876

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112876
Availability details for
Quantity
  • Availability
  • Add to Cart