ENA372573764 Enamine

3-(2-bromophenoxy)pyridazine

  • MDL number MFCD16767534
  • Cas Number : 40580-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112947

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112947
Availability details for
Quantity
  • Availability
  • Add to Cart