ENA372573772 Enamine

2-(2-methoxyethyl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD14661568
  • Cas Number : 1250342-63-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113522

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113522
Availability details for
Quantity
  • Availability
  • Add to Cart