ENA372573777 Enamine

4-amino-N-[2-(diethylamino)ethyl]-2-methylbenzamide

  • MDL number MFCD01679714
  • Cas Number : 30459-64-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113531

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113531
Availability details for
Quantity
  • Availability
  • Add to Cart