ENA372573779 Enamine

1H-1,2,3-triazole-4-carboxamide

  • MDL number MFCD18264276
  • Cas Number : 53897-99-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 256...258
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113005

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113005
Availability details for
Quantity
  • Availability
  • Add to Cart