ENA372573782 Enamine

2-[2,5-dioxo-4-(propan-2-yl)-4-(thiophen-2-yl)imidazolidin-1-yl]acetic acid

  • MDL number MFCD16208421
  • Cas Number : 1274709-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113543

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113543
Availability details for
Quantity
  • Availability
  • Add to Cart