ENA372573783 Enamine

N-[1-(propan-2-yl)piperidin-4-yl]formamide

  • MDL number MFCD21120743
  • Cas Number : 1423025-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113544

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113544
Availability details for
Quantity
  • Availability
  • Add to Cart