ENA372573802 Enamine

1-phenyl-5-(propan-2-yl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD08443788
  • Cas Number : 3191-87-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 136-139
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-113591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113591
Availability details for
Quantity
  • Availability
  • Add to Cart