ENA372573804 Enamine

1-[(cyclopentylcarbamoyl)methyl]-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD06808150
  • Cas Number : 1006454-97-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113595

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113595
Availability details for
Quantity
  • Availability
  • Add to Cart