ENA372573824 Enamine

2-(4-bromophenoxy)-6-chloropyridine-4-carboxylic acid

  • MDL number MFCD06208652
  • Cas Number : 773099-34-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 213...215
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113624

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113624
Availability details for
Quantity
  • Availability
  • Add to Cart