ENA372573829 Enamine

2-[4-cyclopropyl-2,5-dioxo-4-(thiophen-2-yl)imidazolidin-1-yl]acetic acid

  • MDL number MFCD16208422
  • Cas Number : 1270850-79-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113629

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113629
Availability details for
Quantity
  • Availability
  • Add to Cart