ENA372573844 Enamine

1-(3-methoxypyridin-4-yl)methanamine

  • MDL number MFCD11100731
  • Cas Number : 909895-75-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113033

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113033
Availability details for
Quantity
  • Availability
  • Add to Cart