ENA372573867 Enamine

4-{[4-(pyrimidin-2-yl)piperazin-1-yl]methyl}-1,3-thiazol-2-amine

  • MDL number MFCD12460117
  • Cas Number : 1096943-32-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 158...160
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113732

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113732
Availability details for
Quantity
  • Availability
  • Add to Cart