ENA372573897 Enamine

2-(2,2,2-trifluoroethyl)-1,3-thiazole-5-carbaldehyde

  • MDL number MFCD14657114
  • Cas Number : 1250177-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113816

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113816
Availability details for
Quantity
  • Availability
  • Add to Cart