ENA372573901 Enamine

2-[(cyclopentylamino)methyl]benzonitrile hydrochloride

  • MDL number MFCD22421866
  • Cas Number : 1216412-11-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 233...235
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113828

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113828
Availability details for
Quantity
  • Availability
  • Add to Cart