ENA372573916 Enamine

1-[4-(1H-pyrazol-1-yl)phenyl]ethan-1-ol

  • MDL number MFCD08572144
  • Cas Number : 956806-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 92...94
Purity 95
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113895

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113895
Availability details for
Quantity
  • Availability
  • Add to Cart