ENA372573921 Enamine

1-cyclopropylimidazolidin-2-one

  • MDL number MFCD19219176
  • Cas Number : 62491-84-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113900

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113900
Availability details for
Quantity
  • Availability
  • Add to Cart