ENA372573926 Enamine

N-(3-hydroxyphenyl)-6-methylpyridine-2-carboxamide

  • MDL number MFCD11167747
  • Cas Number : 1038243-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 177...179
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113905

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113905
Availability details for
Quantity
  • Availability
  • Add to Cart