ENA372573927 Enamine

tert-butyl 2-amino-3-phenylpropanoate

  • MDL number MFCD12762205
  • Cas Number : 16367-71-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113093

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113093
Availability details for
Quantity
  • Availability
  • Add to Cart