ENA372573931 Enamine

2-methyl-1-[4-(trifluoromethyl)phenyl]propan-1-one

  • MDL number MFCD16309307
  • Cas Number : 64436-58-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113921
Availability details for
Quantity
  • Availability
  • Add to Cart