ENA372573935 Enamine

1-(2,6-dichlorophenyl)-2-methylpropan-1-one

  • MDL number MFCD22421875
  • Cas Number : 1423032-98-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113926

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113926
Availability details for
Quantity
  • Availability
  • Add to Cart