ENA372573936 Enamine

4-(5-oxo-3-phenyl-4,5-dihydro-1H-pyrazol-1-yl)benzonitrile

  • MDL number MFCD13638280
  • Cas Number : 118049-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113927
Availability details for
Quantity
  • Availability
  • Add to Cart