ENA372573944 Enamine

4-methanesulfonylpiperazin-2-one

  • MDL number MFCD17226190
  • Cas Number : 59701-96-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 164...166
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113097

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113097
Availability details for
Quantity
  • Availability
  • Add to Cart