ENA372573945 Enamine

2-amino-N-methylcyclohexane-1-carboxamide

  • MDL number MFCD17219795
  • Cas Number : 1339178-74-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113999

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113999
Availability details for
Quantity
  • Availability
  • Add to Cart