ENA372573948 Enamine

3-iodo-1H-thieno[3,2-c]pyrazole

  • MDL number MFCD16658304
  • Cas Number : 848356-68-3
  • Molecular Weight : 250.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-iodo-1H-thieno[3,2-c]pyrazole
Inchi Code 1S/C5H3IN2S/c6-5-4-3(7-8-5)1-2-9-4/h1-2H,(H,7,8)
Inchi Key VEEHPGKXSQJZSB-UHFFFAOYSA-N

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114003

Safety Information

signalword Warning
pcodes P261,P264,P270,P271,P280,P301+P312,P302+P352,P304+P340,P305+P351+P338,P312,P330,P332+P313,P337+P313,P362,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-114003
pictograms GHS07
hstatements H302,H315,H319,H335
Availability details for
Quantity
  • Availability
  • Add to Cart