ENA372573955 Enamine

1-(3-chlorophenyl)-5-methyl-1H-pyrazol-4-amine

  • MDL number MFCD16042728
  • Cas Number : 1248162-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114014

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114014
Availability details for
Quantity
  • Availability
  • Add to Cart