ENA372573957 Enamine

1-(4-tert-butylphenyl)cyclobutan-1-amine

  • MDL number MFCD12168518
  • Cas Number : 1096853-41-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114016

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114016
Availability details for
Quantity
  • Availability
  • Add to Cart