ENA372573959 Enamine

4-chloro-2-ethoxybenzonitrile

  • MDL number MFCD17290875
  • Cas Number : 548473-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 81...83
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114021

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114021
Availability details for
Quantity
  • Availability
  • Add to Cart