ENA372573963 Enamine

N-cyclopropylpyrrolidin-3-amine

  • MDL number MFCD18278203
  • Cas Number : 1228458-66-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114026

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114026
Availability details for
Quantity
  • Availability
  • Add to Cart