ENA372573966 Enamine

2-(naphthalen-2-yloxy)ethane-1-sulfonyl chloride

  • MDL number MFCD10022959
  • Cas Number : 3383-80-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-112887

Safety Information

msds https://www.enaminestore.com/catalog/EN300-112887
Availability details for
Quantity
  • Availability
  • Add to Cart