ENA372573967 Enamine

2-chloro-4-[(dimethylcarbamoyl)amino]benzoic acid

  • MDL number MFCD14659074
  • Cas Number : 59587-01-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 191...193
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113101

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113101
Availability details for
Quantity
  • Availability
  • Add to Cart