ENA372573971 Enamine

3-amino-N,2,2-trimethylpropanamide

  • MDL number MFCD16117905
  • Cas Number : 756454-05-4
  • Molecular Weight : 130.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form oil
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114112

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114112
Availability details for
Quantity
  • Availability
  • Add to Cart