ENA372573990 Enamine

1-{4-methoxy-3-[(oxan-4-yloxy)methyl]phenyl}methanamine

  • MDL number MFCD15514217
  • Cas Number : 1423032-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114139

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114139
Availability details for
Quantity
  • Availability
  • Add to Cart