ENA372573991 Enamine

1-bromo-4-(but-1-en-2-yl)benzene

  • MDL number MFCD20384655
  • Cas Number : 42427-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113105

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-113105
Availability details for
Quantity
  • Availability
  • Add to Cart