ENA372573993 Enamine

2-hydroxy-3-(4-methoxyphenyl)propanoic acid

  • MDL number MFCD07778404
  • Cas Number : 28030-15-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114147

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114147
Availability details for
Quantity
  • Availability
  • Add to Cart