ENA372573995 Enamine

4-(dimethylamino)-2-fluorobenzonitrile

  • MDL number MFCD22421887
  • Cas Number : 1189353-13-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114150

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114150
Availability details for
Quantity
  • Availability
  • Add to Cart