ENA372574000 Enamine

1-(prop-2-yn-1-yl)imidazolidin-2-one

  • MDL number MFCD14638386
  • Cas Number : 131423-07-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 119...121
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113108

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113108
Availability details for
Quantity
  • Availability
  • Add to Cart