ENA372574010 Enamine

2-(2-amino-5-fluorophenoxy)propanenitrile

  • MDL number MFCD16077309
  • Cas Number : 1250797-65-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 89...91
Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114207

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114207
Availability details for
Quantity
  • Availability
  • Add to Cart