ENA372574018 Enamine

2-(cyclopentylamino)-1-(pyrrolidin-1-yl)ethan-1-one

  • MDL number MFCD11150331
  • Cas Number : 1021131-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114224

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114224
Availability details for
Quantity
  • Availability
  • Add to Cart