ENA372574029 Enamine

[(2-chlorophenyl)methyl]dimethylamine

  • MDL number MFCD04037302
  • Cas Number : 10175-31-2
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114269

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114269
Availability details for
Quantity
  • Availability
  • Add to Cart