ENA372574030 Enamine

4-(1H-pyrrol-1-yl)butanoic acid

  • MDL number MFCD09733323
  • Cas Number : 70686-51-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114270

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114270
Availability details for
Quantity
  • Availability
  • Add to Cart