ENA372574037 Enamine

4-chloro-2-methoxybenzene-1-sulfonamide

  • MDL number MFCD11647455
  • Cas Number : 749253-52-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114279

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114279
Availability details for
Quantity
  • Availability
  • Add to Cart