ENA372574043 Enamine

5-[(4-bromo-1H-pyrazol-1-yl)methyl]furan-3-carboxylic acid

  • MDL number MFCD16702635
  • Cas Number : 1247390-92-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-113128

Safety Information

msds https://www.enaminestore.com/catalog/EN300-113128
Availability details for
Quantity
  • Availability
  • Add to Cart