ENA372574048 Enamine

N-[1-(morpholin-4-yl)propan-2-yl]pyrimidin-4-amine

  • MDL number MFCD14680894
  • Cas Number : 1249287-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-112849

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-112849
Availability details for
Quantity
  • Availability
  • Add to Cart