ENA372574056 Enamine

2-(azetidin-1-yl)ethan-1-amine

  • MDL number MFCD19205720
  • Cas Number : 795299-77-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114312

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114312
Availability details for
Quantity
  • Availability
  • Add to Cart