ENA372574059 Enamine

4-(2-methanesulfonylethyl)aniline

  • MDL number MFCD12779626
  • Cas Number : 596131-24-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 93...95
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114346

Safety Information

signalword Warning
msds https://www.enaminestore.com/catalog/EN300-114346
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart