ENA372574062 Enamine

1-(1-benzylpyrrolidin-3-yl)-4-chloro-1H-pyrazol-5-amine

  • MDL number MFCD16518139
  • Cas Number : 1285592-96-2
  • Molecular Weight : 276.77

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-114352

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-114352
Availability details for
Quantity
  • Availability
  • Add to Cart