ENA372574064 Enamine

2,4-dichloro-6-(2,2,2-trifluoroethoxy)-1,3,5-triazine

  • MDL number MFCD17236948
  • Cas Number : 185677-00-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114354

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114354
Availability details for
Quantity
  • Availability
  • Add to Cart