ENA372574067 Enamine

5-bromopyrazolo[1,5-a]pyrimidine

  • MDL number MFCD11042744
  • Cas Number : 1159981-95-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 131...133
Purity 95.00
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-114365

Safety Information

msds https://www.enaminestore.com/catalog/EN300-114365
Availability details for
Quantity
  • Availability
  • Add to Cart